Skip to Content
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Custom Products
Digital Platforms & Software
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ethyl alcohol, Pure 200 proof
Synonym(s):
Ethyl alcohol, Pure, Ethanol, absolute alcohol, non-denatured ethanol
Linear Formula:
CH
3
CH
2
OH
CAS Number:
64-17-5
Molecular Weight:
46.07
EC Number:
200-578-6
Products
Product Category
solvents (1)
Brand
Sigma-Aldrich (1)
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Quality Segment
Application
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
E7023
Ethyl alcohol, Pure 200 proof,
≥99.45%, for molecular biology, 200 proof, suitable for RNA extraction and DNA extraction
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All