Ethambutol for system suitability
Synonym(s):
Ethambutol dihydrochloride, 2,2′-(1,2-Ethanediyldiimino)bis-1-butanol dihydrochloride, Emb
Empirical Formula (Hill Notation):
C10H24N2O2 · 2HCl
CAS Number:
Molecular Weight:
277.23
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing