Skip to Content
Merck
vibrant-m

Entecavir Related Compound A

vibrant-m
Synonym(s):
9-[(1S,3R,4S)-4-Dimethylphenylsilyl-3-(benzyloxymethyl)-2-methylenecyclopentyl]guanine, 2-Amino-9-[(1S,3R,4S)-3-[(benzyloxy)methyl]-4-[dimethyl(phenyl)silyl]-2-methylidenecyclopentyl]-1,9-dihydro-6H-purin-6-one, 2-Amino-9-[(1S,3R,4S)-4-(dimethylphenylsilyl)-2-methylene-3-[(phenylmethoxy)methyl]cyclopentyl]-1,9-dihydro-6H-purin-6-one
Empirical Formula (Hill Notation):
C27H31N5O2Si
CAS Number:
Molecular Weight:
485.65

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1