
E7820-C9-NH2 hydrochloride

Empirical Formula (Hill Notation):
C26H33N5O3S · xHCl
Molecular Weight:
495.64 (free base basis)
Products
Product Category
Brand
Formula Weight
Quality Segment
Functional Group
Reactivity
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing