Skip to Content
Merck
vibrant-m

Diazepam Related Compound B

vibrant-m
Synonym(s):
3-Amino-6-chloro-1-methyl-4-phenylcarbostyril, 3-Amino-6-chloro-1-methyl-4-phenyl-2(1H)-quinolinone
Empirical Formula (Hill Notation):
C16H13ClN2O
CAS Number:
Molecular Weight:
284.74

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade