Skip to Content
Merck
cis-Stilbene oxide 97%

cis-Stilbene oxide

cis-Stilbene oxide 97%
Synonym(s):
cis-2,3-Diphenyloxirane
Empirical Formula (Hill Notation):
C14H12O
CAS Number:
Molecular Weight:
196.24

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing