Skip to Content
Merck
Cinnarizine European Pharmacopoeia (EP) Reference Standard

Cinnarizine

Cinnarizine European Pharmacopoeia (EP) Reference Standard
Synonym(s):
1-trans-Cinnamyl-4-diphenylmethylpiperazine, Cinnarizine
Empirical Formula (Hill Notation):
C26H28N2
CAS Number:
Molecular Weight:
368.51

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade