Carbidopa
Synonym(s):
(S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropionic acid monohydrate
Empirical Formula (Hill Notation):
C10H14N2O4 · H2O
CAS Number:
Molecular Weight:
244.24
Products
Shipping
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing