
Calcium folinate for system suitability

Synonym(s):
Folinic acid calcium salt hydrate, 5-Formyl-5,6,7,8-tetrahydrofolic acid calcium salt, 5-Formyl-5,6,7,8-tetrahydropteroyl-L-glutamic acid calcium salt, 5-HCO-H4PteGlu, Calcium folinate, Citrovorum factor calcium salt, Leucovorin calcium salt
Empirical Formula (Hill Notation):
C20H21CaN7O7 · xH2O
CAS Number:
Molecular Weight:
511.50 (anhydrous basis)
Products
Product Category
Biological Source
Formula Weight
Manufacturer
Physical Form
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing