Skip to Content
Merck
Bromocriptine mesilate British Pharmacopoeia (BP) Reference Standard

Bromocriptine mesilate

Bromocriptine mesilate British Pharmacopoeia (BP) Reference Standard
Synonym(s):
2-Bromo-α-ergocryptine methanesulfonate salt, (+)-2-Bromo-12′-hydroxy-2′-(1-methylethyl)-5′-(2-methylpropyl)ergotaman-3′,6′-18-trione methanesulfonate salt, (+)-Bromocriptine methanesulfonate salt, Bromocriptine mesylate salt
Empirical Formula (Hill Notation):
C32H40BrN5O5 · CH4SO3
CAS Number:
Molecular Weight:
750.70

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing