Brilliant cresyl blue zinc chloride double salt
Synonym(s):
Brilliant cresyl blue zinc chloride double salt
Linear Formula:
(C17H20N3O)2 · ZnCl4
CAS Number:
Molecular Weight:
771.92
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing