Bombesin acetate salt hydrate
Synonym(s):
pGlu-Gln-Arg-Leu-Gly-Asn-Gln-Trp-Ala-Val-Gly-His-Leu-Met-NH2
Empirical Formula (Hill Notation):
C71H110N24O18S · xC2H4O2 · yH2O
Molecular Weight:
1619.85 (anhydrous free base basis)
Products
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing