![Boc-Lys(2-Cl-Z)-OH (cryst) Novabiochem®](/deepweb/assets/sigmaaldrich/product/images/122/093/6dc1f758-683f-4778-953f-585aa99605d8/640/6dc1f758-683f-4778-953f-585aa99605d8.jpg)
Boc-Lys(2-Cl-Z)-OH
![Boc-Lys(2-Cl-Z)-OH (cryst) Novabiochem®](/deepweb/assets/sigmaaldrich/product/images/122/093/6dc1f758-683f-4778-953f-585aa99605d8/640/6dc1f758-683f-4778-953f-585aa99605d8.jpg)
Synonym(s):
Boc-Lys(2-Cl-Z)-OH
Empirical Formula (Hill Notation):
C19H27ClN2O6
CAS Number:
Molecular Weight:
414.88
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing