
Benzalkonium bromide (C12)

Synonym(s):
Benzyldodecyldimethylammonium bromide, Benzyldimethyldodecylammonium bromide
Linear Formula:
CH3(CH2)11N(Br)(CH3)2CH2C6H5
CAS Number:
Molecular Weight:
384.44
Products
Shipping
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing