Skip to Content
Merck
Azlocillin sodium salt

Azlocillin sodium salt

Azlocillin sodium salt
Synonym(s):
D-α-([Imidazolidin-2-on-1-yl]carbonylamino)benzylpenicillin
Linear Formula:
C20H22N5O6SNa
CAS Number:
Molecular Weight:
483.47
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Activity Spectrum
Mode of Action
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing