Skip to Content
Merck
Astaxanthin ≥97% (HPLC)

Astaxanthin

Astaxanthin ≥97% (HPLC)
Synonym(s):
(3S,3′S)-3,3′-dihydroxy-β,β-Carotene-4,4′-dione, trans-Astaxanthin
Empirical Formula (Hill Notation):
C40H52O4
CAS Number:
Molecular Weight:
596.84

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing