![Aristolocholic acid for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/734/489/e049e551-869e-43f7-8542-e58f9b742247/640/e049e551-869e-43f7-8542-e58f9b742247.png)
Aristolocholic acid for system suitability
![Aristolocholic acid for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/734/489/e049e551-869e-43f7-8542-e58f9b742247/640/e049e551-869e-43f7-8542-e58f9b742247.png)
Synonym(s):
Aristolochic acid I sodium salt
Empirical Formula (Hill Notation):
C17H10NNaO7
CAS Number:
Molecular Weight:
363.25
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing