![Anti-Vinculin antibody produced in rabbit affinity isolated antibody, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/199/319/e99863c8-efa7-4e0b-a099-cfe7c5890d5c/640/e99863c8-efa7-4e0b-a099-cfe7c5890d5c.jpg)
Anti-Vinculin antibody produced in rabbit
![Anti-Vinculin antibody produced in rabbit affinity isolated antibody, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/199/319/e99863c8-efa7-4e0b-a099-cfe7c5890d5c/640/e99863c8-efa7-4e0b-a099-cfe7c5890d5c.jpg)
Synonym(s):
Metavinculin, VCL, VINC
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2