![Anti-TST antibody produced in rabbit affinity isolated antibody](/deepweb/assets/sigmaaldrich/product/images/273/178/31376e2f-e973-46ee-a280-1dd3338c6b6f/640/31376e2f-e973-46ee-a280-1dd3338c6b6f.jpg)
Anti-TST antibody produced in rabbit
![Anti-TST antibody produced in rabbit affinity isolated antibody](/deepweb/assets/sigmaaldrich/product/images/273/178/31376e2f-e973-46ee-a280-1dd3338c6b6f/640/31376e2f-e973-46ee-a280-1dd3338c6b6f.jpg)
Synonym(s):
Anti-Rhodanese antibody produced in rabbit, Anti-Thiosulfate sulfurtransferase antibody produced in rabbit, Anti-MGC19578, Anti-RDS, Anti-Thiosulfate sulfurtransferase (rhodanese)
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing