Anti-Sheep Serum antibody produced in rabbit
Synonym(s):
Anti-Serum, sheep
Products
Product Category
Brand
Biological Source
Clonality
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing