Skip to Content
Merck
Anti-PAK1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-PAK1 antibody produced in rabbit

Anti-PAK1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-Alpha-PAK, Anti-PAK-1, Anti-Serine/threonine-protein kinase PAK 1, Anti-p21-activated kinase 1, Anti-p65-PAK, α-PAK, PAK-1, Serine/threonine-protein kinase PAK 1, p21-activated kinase 1, p65-PAK, γ-PAK, P21-activated kinase 2, PAK 2, PAK-2, PAK65

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique