Anti-NDUFAF6 antibody produced in rabbit
Synonym(s):
Anti-MGC40214, Anti-chromosome 8 open reading frame 38
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Product Line
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1