Anti-MAPK3 antibody produced in rabbit
Synonym(s):
Anti-ERK-1 antibody produced in rabbit, Anti-ERT2 antibody produced in rabbit, Anti-Extracellular signal-regulated kinase 1 antibody produced in rabbit, Anti-Insulin-stimulated MAP2 kinase antibody produced in rabbit, Anti-MAP kinase 1 antibody produced in rabbit, Anti-MAPK 1 antibody produced in rabbit, Anti-Microtubule-associated protein 2 kinase antibody produced in rabbit, Anti-Mitogen-activated protein kinase 3 antibody produced in rabbit, Anti-p44-ERK1 antibody produced in rabbit, Anti-p44-MAPK antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing