Anti-gapdh antibody produced in rabbit
Synonym(s):
cb609, gapd, gapdh, mg:bb02e05
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Conjugate
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing