![Amoxicillin Related Compound F Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/361/518/92e6e915-294d-4b48-a504-360f95619546/640/92e6e915-294d-4b48-a504-360f95619546.jpg)
Amoxicillin Related Compound F
![Amoxicillin Related Compound F Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/361/518/92e6e915-294d-4b48-a504-360f95619546/640/92e6e915-294d-4b48-a504-360f95619546.jpg)
Synonym(s):
3-(4-Hydroxyphenyl)pyrazin-2-ol
Empirical Formula (Hill Notation):
C10H8N2O2
CAS Number:
Molecular Weight:
188.18
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2