Skip to Content
Merck
Aluminum sulfate hexadecahydrate SAJ special grade

Aluminum sulfate hexadecahydrate

Aluminum sulfate hexadecahydrate SAJ special grade
Linear Formula:
Al2(SO4)3 · 16H2O
CAS Number:
Molecular Weight:
630.40

Products

Brand
Formula Weight
Physical Form
Showing 1-2 of 2