![Acetyl Simvastatin certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/303/870/2d174849-6523-4cec-b441-bc41a82f15e9/640/2d174849-6523-4cec-b441-bc41a82f15e9.jpg)
Acetyl simvastatin
![Acetyl Simvastatin certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/303/870/2d174849-6523-4cec-b441-bc41a82f15e9/640/2d174849-6523-4cec-b441-bc41a82f15e9.jpg)
Synonym(s):
4′-Acetylsimvastatin, (1S,3R,7S,8S,8aR)-8-{2-[(2R,4R)-4-(Acetoxy)-6-oxotetrahydro-2H-pyran-2-yl]ethyl}-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2,2-dimethylbutanoate
Empirical Formula (Hill Notation):
C27H40O6
CAS Number:
Molecular Weight:
460.60
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2