Skip to Content
Merck
8-Chlorotheophylline United States Pharmacopeia (USP) Reference Standard

8-Chlorotheophylline

8-Chlorotheophylline United States Pharmacopeia (USP) Reference Standard
Synonym(s):
8-Chloro-1,3-dimethyl-2,6(1H,3H)-purinedione
Empirical Formula (Hill Notation):
C7H7ClN4O2
CAS Number:
Molecular Weight:
214.61

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing