Skip to Content
JP
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Custom Products
Digital Platforms & Software
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
7-METHOXY-2,2-DIPHENYL-BENZO(1,3)DIOXOLE-5-CARBOXYLIC ACID METHYL ESTER
Linear Formula:
C
22
H
18
O
5
CAS Number:
102706-14-9
Molecular Weight:
362.386
Products
Formula Weight
Special Grade
Aldrich
CPR
(2)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
S105392
7-METHOXY-2,2-DIPHENYL-BENZO(1,3)DIOXOLE-5-CARBOXYLIC ACID METHYL ESTER,
Aldrich
CPR
Expand
View Pricing
S13022
7-METHOXY-2,2-DIPHENYL-BENZO(1,3)DIOXOLE-5-CARBOXYLIC ACID METHYL ESTER,
Aldrich
CPR
Expand
View Pricing