
6-(p-Toluidino)-2-naphthalenesulfonyl chloride

Synonym(s):
2,6-TNS-Cl, 6-(4-Methylanilino)naphthalene-2-sulfonylchloride, 6-[(4-methylphenyl)amino]-2-naphthalenesulfonylchloride
Empirical Formula (Hill Notation):
C17H14ClNO2S
CAS Number:
Molecular Weight:
331.82
Products
Brand
Formula Weight
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing