![5-([4,6-Dichlorotriazin-2-yl]amino)fluorescein hydrochloride ≥90% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/271/656/5f185b68-9254-440f-8211-1f4fbe1fee16/640/5f185b68-9254-440f-8211-1f4fbe1fee16.png)
5-([4,6-Dichlorotriazin-2-yl]amino)fluorescein hydrochloride
![5-([4,6-Dichlorotriazin-2-yl]amino)fluorescein hydrochloride ≥90% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/271/656/5f185b68-9254-440f-8211-1f4fbe1fee16/640/5f185b68-9254-440f-8211-1f4fbe1fee16.png)
Synonym(s):
DTAF
Empirical Formula (Hill Notation):
C23H12Cl2N4O5 Ā· HCl
CAS Number:
Molecular Weight:
531.73
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing