
(4-Methylbenzyl)hydrazine hydrochloride

Empirical Formula (Hill Notation):
C8H13ClN2
CAS Number:
Molecular Weight:
172.66
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing