4-Methoxy-o-phenylenediamine dihydrochloride
Synonym(s):
3,4-Diaminoanisole dihydrochloride
Linear Formula:
CH3OC6H3(NH2)2·2HCl
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing