4-[2-[Benzyl(tert-butyl)amino]-1-hydroxyethyl]-2-(hydroxymethyl)phenol
Synonym(s):
α1-[[(1,1-Dimethylethyl)(phenylmethyl)amino]methyl]-4-hydroxy-1,3-benzenedimethanol, (1RS)-2-[Benzyl(1,1-dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanol
Empirical Formula (Hill Notation):
C20H27NO3
CAS Number:
Molecular Weight:
329.43
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1