Skip to Content
Merck
vibrant-m

3-(Piperazin-1-yl)benzo[d]isothiazole monohydrochloride

vibrant-m
Empirical Formula (Hill Notation):
C11H13N3S · HCl
CAS Number:
Molecular Weight:
255.77

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade