Skip to Content
Merck
1-Piperazinecarboxylic acid, 4-(4-piperidinyl)-, 1,1-dimethylethyl ester ≥98%

1-Piperazinecarboxylic acid, 4-(4-piperidinyl)-, 1,1-dimethylethyl ester

1-Piperazinecarboxylic acid, 4-(4-piperidinyl)-, 1,1-dimethylethyl ester ≥98%
Synonym(s):
1-Boc-4-(piperidin-4-yl)-piperazine
Empirical Formula (Hill Notation):
C14H27N3O2
CAS Number:
Molecular Weight:
269.38

Products

Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1