Skip to Content
Merck
1-Phenyldodecane analytical standard

1-Phenyldodecane

1-Phenyldodecane analytical standard
Synonym(s):
Dodecylbenzene, 1-Phenyldodecane
Linear Formula:
C6H5(CH2)11CH3
CAS Number:
Molecular Weight:
246.43
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing