Skip to Content
Merck
1,3,2-Dioxathiolane 2,2-dioxide 98%

1,3,2-Dioxathiolane 2,2-dioxide

1,3,2-Dioxathiolane 2,2-dioxide 98%
Empirical Formula (Hill Notation):
C2H4O4S
CAS Number:
Molecular Weight:
124.12

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing