Skip to Content
Merck
vibrant-m

1,1′-Ethylidenebistryptophan

vibrant-m
Synonym(s):
1,1′-Ethylidenebis-(L-tryptophan), EBT
Empirical Formula (Hill Notation):
C24H26N4O4
CAS Number:
Molecular Weight:
434.49

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1