Key Documents
29112-U
Supelco
Inter-Seal bonded caps (inseparable caps and liner)
black cap, PTFE/silicone septa, thread for 10-425
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
PTFE/silicone septa
black cap
packaging
pkg of 100 ea
fitting
thread for 10-425
General description
Inter-Seal bonded caps eliminate expensive and time consuming hand assembly while preventing liner fallout. They are made by a bonding process that bonds PTFE/silicone into polypropylene closures without using adhesives. The bonding properly seats and secures the septa within the vial opening. This integrated one piece system withstands multiple injections, making it very cost effective.
Benefits include:
  • Significant bond strength
  • Withstands multiple injections
  • Resistant to coring
  • Excellent resealability
  • Broad chemical resistance
Legal Information
Inter-Seal is a trademark of Integrated Liner Technologies, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)