Skip to Content
Merck
All Photos(1)

Documents

SAB1402732

Sigma-Aldrich

Monoclonal Anti-COL9A1 antibody produced in mouse

clone 3H1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

DJ149L1.1.2, EDM6, FLJ40263, MED

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3H1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~62.19 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COL9A1(1297)

General description

This gene encodes one of the three alpha chains of type IX collagen, which is a minor (5-20%) collagen component of hyaline cartilage. Type IX collagen is usually found in tissues containing type II collagen, a fibrillar collagen. Studies in knockout mice have shown that synthesis of the alpha 1 chain is essential for assembly of type IX collagen molecules, a heterotrimeric molecule, and that lack of type IX collagen is associated with early onset osteoarthritis. Mutations in this gene are associated with osteoarthritis in humans, with multiple epiphyseal dysplasia, 6, a form of chondrodysplasia, and with Stickler syndrome, a disease characterized by ophthalmic, orofacial, articular, and auditory defects. Two transcript variants that encode different isoforms have been identified for this gene. (provided by RefSeq)

Immunogen

COL9A1 (AAH15409, 1 a.a. ~ 328 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MKTCWKIPVFFFVCSFLEPWASAAVKRRPRFPVNSNSNGGNELCPKIRIGQDDLPGFDLISQFQVDKAASRRAIQRVVGSATLQVAYKLGNNVDFRIPTRNLYPSGLPEEYSFLTTFRMTGSTLKKNWNIWQIQDSSGKEQVGIKINGQTQSVVFSYKGLDGSLQTAAFSNLSSLFDSQWHKIMIGVERSSATLFVDCNRIESLPIKPRGPIDIDGFAVLGKLADNPQVSVPFELQWMLIHCDPLRPRRETCHELPARITPSQTTDERGPPGEQGPPGPPGPPGVPGIDGIDGDRGPKGPPGPPGPAGEPGKPGAPGKPGTPGADTSP

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service