Skip to Content
Merck
All Photos(1)

Key Documents

75898

Sigma-Aldrich

Dimethyl 2-oxoheptylphosphonate

technical, ~90% (GC)

Synonym(s):

(2-Oxoheptyl)phosphonic acid dimethyl ester, Dimethyl (hexanoylmethyl)phosphonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4COCH2P(O)(OCH3)2
CAS Number:
Molecular Weight:
222.22
Beilstein:
1946305
EC Number:
MDL number:
UNSPSC Code:
12352000

grade

technical

Assay

~90% (GC)

refractive index

n20/D 1.444 (lit.)
n20/D 1.444

bp

109 °C/0.4 mmHg (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

CCCCCC(=O)CP(=O)(OC)OC

InChI

1S/C9H19O4P/c1-4-5-6-7-9(10)8-14(11,12-2)13-3/h4-8H2,1-3H3

InChI key

LQZCYXCHWNQBKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Beirut reaction for the preparation of phosphonylated quinoxaline dioxides
  • Preparation of pyrrolomorphinan derivatives as κ opioid agonists via cyclocondensation, Vilsmeier-Haack formylation, Horner-Wadsworth-Emmons reaction, and Kocienski-modified Julia olefination from naltrexone methyl ether
  • Synthesis of C-glycosides amphiphiles via solvent-free Horner-Wadsworth-Emmons reaction with unprotected sugars
  • Preparation of specific inhibitors of endocannabinoid biosynthesis

Other Notes

Wittig-Horner reagent for the synthesis of 1-substituted 1-octen-3-ones from aldehydes, especially for introducing the ω-chain in prostaglandins and thromboxanes

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Steliou et al.
The Journal of Organic Chemistry, 54, 5128-5128 (1989)
S. Cook et al.
Journal of the Chemical Society. Perkin Transactions 1, 1825-1825 (1987)
R.K. Taylor et al.
Prostaglandins and Thromboxanes (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service