Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

32668

Supelco

Nabam-d4 hexahydrate

PESTANAL®, analytical standard

Synonym(s):

Disodium ethylene-d4-bis(dithiocarbamate) hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4D4H2N2Na2S4 · 6H2O
Molecular Weight:
368.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

neat

mass shift

M+4

SMILES string

O.O.O.O.O.O.[Na+].[Na+].[2H]C([2H])(NC([S-])=S)C([2H])([2H])NC([S-])=S

InChI

1S/C4H8N2S4.2Na.6H2O/c7-3(8)5-1-2-6-4(9)10;;;;;;;;/h1-2H2,(H2,5,7,8)(H2,6,9,10);;;6*1H2/q;2*+1;;;;;;/p-2/i1D2,2D2;;;;;;;;

InChI key

JPLBPVJXWHMSLP-KJGMLHTRSA-L

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

32668-VAR:
32668-BULK:
32668-10MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service