ENAH5802EDE2
Enamine
2-(dimethylamino)benzene-1,4-diol
About This Item
mp
93-95
Iupac Name
2-(dimethylamino)benzene-1,4-diol
Inchi Code
1S/C8H11NO2/c1-9(2)7-5-6(10)3-4-8(7)11/h3-5,10-11H,1-2H3
InChI key
UWUNWZJAUUXNBQ-UHFFFAOYSA-N
1 of 4
This Item | 724939 | 367095 | 726842 |
---|---|---|---|
100 | 100 | 100 | 100 |
n20/D 1.5347 | n20/D 1.528 | n20/D 1.516 (lit.) | - |
1.0190 g/mL at 25 °C | 1.002 g/mL at 25 °C | 0.979 g/mL at 25 °C (lit.) | 1.024 g/mL at 20 °C (lit.) |
liquid | liquid | liquid | liquid |
amine | amine | - | amine |
Purity
Country of Origin
Product Link
Melting Point
MSDS
https://www.enaminestore.com/catalog/EN300-7442044
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service