Skip to Content
Merck
All Photos(1)

Key Documents

COM448660433

Combi-Blocks, Inc.

Acetaldehyde butyl 2,2,2-trifluoroethyl acetal

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
¥12,200
5 G
¥19,200
25 G
¥39,100
100 G
¥103,400

Vendor SKU: QC-7195-1G

¥12,200


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
¥12,200
5 G
¥19,200
25 G
¥39,100
100 G
¥103,400

About This Item

CAS Number:
Molecular Weight:
200.2
MDL number:

Vendor SKU: QC-7195-1G

¥12,200


Ships in 14 days - from Aldrich Partner.

Iupac Name

1-[1-(2,2,2-trifluoroethoxy)ethoxy]butane

Inchi Code

1S/C8H15F3O2/c1-3-4-5-12-7(2)13-6-8(9,10)11/h7H,3-6H2,1-2H3

InChI key

DIKMXGDAKSQQSJ-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QC-7195

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QC-7195


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service