Skip to Content
Merck
All Photos(1)

Key Documents

CH6371387359

Chem Impex International, Inc.

1-(4-(4-Nitrophenoxy)phenyl)ethanone

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
¥21,200
5 G
¥65,300
25 G
¥212,300

Vendor SKU: 25654-1G

¥21,200


Ships in 12 days - from Aldrich Partner.


Select a Size

Change View
1 G
¥21,200
5 G
¥65,300
25 G
¥212,300

About This Item

CAS Number:
Molecular Weight:
257.25
MDL number:

Vendor SKU: 25654-1G

¥21,200


Ships in 12 days - from Aldrich Partner.

mp

75-81 C

Iupac Name

1-[4-(4-nitrophenoxy)phenyl]ethanone

Inchi Code

1S/C14H11NO4/c1-10(16)11-2-6-13(7-3-11)19-14-8-4-12(5-9-14)15(17)18/h2-9H,1H3

InChI key

IRWXEFXORSDYQA-UHFFFAOYSA-N

storage temp.

Store at 0-8C

Purity

98%

Country of Origin

US

Shipping Temperature

RT

Physical Form

Pale yellow crystals

Product Link

https://www.chemimpex.com/product/productinfo/1-4-4-nitrophenoxy-phenyl-ethanone/16183?cid=0

Melting Point

75-81 C

MSDS

https://www.chemimpex.com/product/productinfo/1-4-4-nitrophenoxy-phenyl-ethanone/16183?cid=0


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service