CH4424847944
ChemBridge Corporation
[trans-4-methoxytetrahydro-3-furanyl]amine hydrochloride
Select a Size
Vendor SKU: 4031140-250MG
¥27,900
Availability
Select a Size
About This Item
Vendor SKU: 4031140-250MG
¥27,900
Availability
Iupac Name
(3S,4R)-4-methoxytetrahydrofuran-3-amine hydrochloride
Inchi Code
1S/C5H11NO2.ClH/c1-7-5-3-8-2-4(5)6;/h4-5H,2-3,6H2,1H3;1H/t4-,5-;/m0./s1
InChI key
KDANNXAXAYGSFR-FHAQVOQBSA-N
storage temp.
Salt Data
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
MSDS
https://www.hit2lead.com/sds/4031140_sds.pdf
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service