Skip to Content
Merck
All Photos(1)

Key Documents

AMBH97B9F38C

Ambeed, Inc.

2-(1,3-Dioxolan-2-yl)ethyltriphenylphosphonium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
¥10,200
25 G
¥17,700
100 G
¥42,200

Vendor SKU: A566804-5G

¥10,200


Ships in 12 days - from Aldrich Partner.


Select a Size

Change View
5 G
¥10,200
25 G
¥17,700
100 G
¥42,200

About This Item

CAS Number:
Molecular Weight:
443.32
MDL number:

Vendor SKU: A566804-5G

¥10,200


Ships in 12 days - from Aldrich Partner.

Iupac Name

[2-(1,3-dioxolan-2-yl)ethyl](triphenyl)phosphonium bromide

Inchi Code

1S/C23H24O2P.BrH/c1-4-10-20(11-5-1)26(21-12-6-2-7-13-21,22-14-8-3-9-15-22)19-16-23-24-17-18-25-23;/h1-15,23H,16-19H2;1H/q+1;/p-1

InChI key

ZCJKBPSRKLHANV-UHFFFAOYSA-M

storage temp.

Inert atmosphere,2-8C

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/86608-70-0.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.ambeed.com/products/86608-70-0.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service