Skip to Content
Merck
All Photos(1)

Key Documents

AMBH9614FA44

Ambeed, Inc.

Allyl(chloromethyl)dimethylsilane

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
¥11,300
5 G
¥23,500
25 G
¥65,600

Vendor SKU: A634612-1G

¥11,300


Ships in 19 days - from Aldrich Partner.


Select a Size

Change View
1 G
¥11,300
5 G
¥23,500
25 G
¥65,600

About This Item

CAS Number:
Molecular Weight:
148.71
MDL number:

Vendor SKU: A634612-1G

¥11,300


Ships in 19 days - from Aldrich Partner.

Iupac Name

allyl(chloromethyl)dimethylsilane

Inchi Code

1S/C6H13ClSi/c1-4-5-8(2,3)6-7/h4H,1,5-6H2,2-3H3

InChI key

UHHLOVCFFWGSMO-UHFFFAOYSA-N

storage temp.

Inert atmosphere,Room Temperature

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.ambeed.com/products/33558-75-7.html

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.ambeed.com/products/33558-75-7.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service