Skip to Content
Merck
All Photos(1)

Key Documents

AMBH9614F059

Ambeed, Inc.

Pent-4-yne-1-sulfonyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
¥22,100
100 MG
¥32,100
250 MG
¥60,600
1 G
¥168,100

Vendor SKU: A1064707-50MG

¥22,100


Ships in 19 days - from Aldrich Partner.


Select a Size

Change View
50 MG
¥22,100
100 MG
¥32,100
250 MG
¥60,600
1 G
¥168,100

About This Item

CAS Number:
Molecular Weight:
166.63
MDL number:

Vendor SKU: A1064707-50MG

¥22,100


Ships in 19 days - from Aldrich Partner.

Iupac Name

4-pentyne-1-sulfonyl chloride

Inchi Code

1S/C5H7ClO2S/c1-2-3-4-5-9(6,7)8/h1H,3-5H2

InChI key

NYFAHXOSIXLIKV-UHFFFAOYSA-N

storage temp.

Inert atmosphere,Store in freezer, under -20C

Purity

95%

Country of Origin

CN

Shipping Temperature

Cold-chain transportation

Physical Form

liquid

Product Link

https://www.ambeed.com/products/67448-64-0.html

Synonym

Pent-4-yne-1-sulfonyl chloride

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service